accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 854689-07-9

IUPAC Name: 4-hydrazinylbenzene-1-sulfonic acid hydrate
Synonyms: 4-hydrazinylbenzenesulfonic acid hydrate
Molecular Weight (g/mol): 206.22
Molecular Formula: C6H10N2O4S
InChi Key: OSOMMFVBTYFMBI-UHFFFAOYSA-N
SMILES: O.NNC1=CC=C(C=C1)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)