accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NNPOH3CH3CCH3

CAS RN 854929-38-7

IUPAC Name: 1,3-di-tert-butyl-2-oxo-1,3,2λ⁵-diazaphospholidin-2-ylium
Synonyms: 1,3-di-tert-butyl-2-oxo-1,3,2λ⁵-diazaphospholidin-2-ylium
Molecular Weight (g/mol): 217.27
Molecular Formula: C10H22N2OP
InChi Key: RBABEMWOZLGIIE-UHFFFAOYSA-N
SMILES: CC(C)(C)N1CCN([P+]1=O)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)