accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 854952-58-2

IUPAC Name: (9-phenyl-9H-carbazol-3-yl)boronic acid
Synonyms: 9-phenylcarbazol-3-ylboronic acid
Molecular Weight (g/mol): 287.13
Molecular Formula: C18H14BNO2
InChi Key: JWJQEUDGBZMPAX-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)