CAS RN 85559-46-2
IUPAC Name:
4-[3-(trifluoromethyl)-3H-diazirin-3-yl]benzoic acid
Synonyms:
4-[3-(trifluoromethyl)diazirin-3-yl]benzoic acid
Molecular Weight (g/mol):
230.15
Molecular Formula:
C9H5F3N2O2
InChi Key:
CZPAJVBVULSLGG-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(C=C1)C1(N=N1)C(F)(F)F
1
–
1
of
1
results
1
4-[3-(Trifluoromethyl)-3H-diazirin-3-yl]benzoic Acid 98.0+%, TCI America™
CAS: 85559-46-2 Molecular Formula: C9H5F3N2O2 Molecular Weight (g/mol): 230.146 MDL Number: MFCD00236809 InChI Key: CZPAJVBVULSLGG-UHFFFAOYSA-N PubChem CID: 128718 IUPAC Name: 4-[3-(trifluoromethyl)diazirin-3-yl]benzoic acid SMILES: C1=CC(=CC=C1C(=O)O)C2(N=N2)C(F)(F)F