accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85609-03-6

IUPAC Name: 1-tert-butyl-4-(phenylsulfanyl)benzene
Synonyms: 1-tert-butyl-4-(phenylsulfanyl)benzene
Molecular Weight (g/mol): 242.38
Molecular Formula: C16H18S
InChi Key: KGRAEJDVHXHGSQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(SC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results