CAS RN 85618-21-9
1-S-Octyl-β-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals
CAS: 85618-21-9 Molecular Formula: C14H28O5S Molecular Weight (g/mol): 308.44 MDL Number: MFCD00012189 InChI Key: CGVLVOOFCGWBCS-DGTMBMJNSA-N Synonym: octyl thioglucopyranoside,octyl-i 2-d-1-thioglucopyranoside PubChem CID: 53720324 IUPAC Name: (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol SMILES: CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
MilliporeSigma™ n-Octyl-β-D-Thioglucopyranoside, ULTROL™ Grade, Calbiochem™,
CAS: 85618-21-9 Molecular Formula: C14H28O5S Molecular Weight (g/mol): 308.433 InChI Key: CGVLVOOFCGWBCS-DGTMBMJNSA-N Synonym: OSGP PubChem CID: 53720324 IUPAC Name: (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol SMILES: CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
n-Octyl-beta-D-thioglucopyranoside, 98+%
CAS: 85618-21-9 Molecular Formula: C14H28O5S Molecular Weight (g/mol): 308.433 MDL Number: MFCD00012189 InChI Key: CGVLVOOFCGWBCS-DGTMBMJNSA-N Synonym: octyl thioglucopyranoside,octyl-i 2-d-1-thioglucopyranoside PubChem CID: 53720324 IUPAC Name: (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol SMILES: CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O
n-Octyl-beta-D-thioglucopyranoside, Electrophoresis Grade, 98+%
CAS: 85618-21-9 Molecular Formula: C14H28O5S Molecular Weight (g/mol): 308.433 MDL Number: MFCD00012189 InChI Key: CGVLVOOFCGWBCS-DGTMBMJNSA-N Synonym: octyl thioglucopyranoside,octyl-i 2-d-1-thioglucopyranoside PubChem CID: 53720324 IUPAC Name: (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol SMILES: CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O