CAS RN 85642-11-1
CAS RN 85642-11-1
IUPAC Name:
5-(1,3-benzothiazol-2-yl)-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Synonyms:
5-(1,3-benzothiazol-2-yl)-3-oxa-13-azatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1,5,7,9(17)-tetraen-4-one
Molecular Weight (g/mol):
374.46
Molecular Formula:
C22H18N2O2S
InChi Key:
WYZWJLZUSHFFOR-UHFFFAOYSA-N
SMILES:
O=C1OC2=C3CCCN4CCCC(C=C2C=C1C1=NC2=CC=CC=C2S1)=C34
1
–
1
of
1
results
1
Coumarin 545 97.0+%, TCI America™
CAS: 85642-11-1 Molecular Formula: C22H18N2O2S Molecular Weight (g/mol): 374.458 InChI Key: WYZWJLZUSHFFOR-UHFFFAOYSA-N Synonym: 10-(2-Benzothiazolyl)-2,3,6,7-tetrahydro-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one PubChem CID: 5237252 SMILES: C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C5=NC6=CC=CC=C6S5)CCCN3C1