accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONH2NHOOCH3CH3H3C

CAS RN 85642-13-3

IUPAC Name: tert-butyl N-(1-carbamoylethyl)carbamate
Synonyms: tert-butyl N-(1-carbamoylethyl)carbamate
Molecular Weight (g/mol): 188.23
Molecular Formula: C8H16N2O3
InChi Key: GZKKSKKIABUUCX-UHFFFAOYNA-N
SMILES: CC(NC(=O)OC(C)(C)C)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)