accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CNBr

CAS RN 856422-39-4

IUPAC Name: 2-bromo-9-(2-ethylhexyl)-9H-carbazole
Synonyms: 2-bromo-9-(2-ethylhexyl)carbazole
Molecular Weight (g/mol): 358.32
Molecular Formula: C20H24BrN
InChi Key: KUHJVARVXWNMAR-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1C2=C(C=CC=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results