accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85694-31-1

IUPAC Name: methyl (2E)-4,4,4-trifluorobut-2-enoate
Synonyms: methyl (2E)-4,4,4-trifluorobut-2-enoate
Molecular Weight (g/mol): 154.09
Molecular Formula: C5H5F3O2
InChi Key: DMMZYYLXAGRBDO-NSCUHMNNSA-N
SMILES: COC(=O)\C=C\C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results