CAS RN 857064-38-1
CAS RN 857064-38-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ STAT3 Inhibitor III, WP1066
STAT3 Inhibitor III, WP1066, CAS 857064-38-1, is a cell-permeable AG490 tyrphostin analog that acts as a STAT3 pathway inhibitor. A potent anti-tumor agent.