accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85720-62-3

IUPAC Name: 2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiol-2-one
Synonyms: 5H-[1,3]dithiolo[4,5-d][1,3]dithiol-2-one
Molecular Weight (g/mol): 194.3
Molecular Formula: C4H2OS4
InChi Key: VBZVKFOMXXSGFB-UHFFFAOYSA-N
SMILES: O=C1SC2=C(SCS2)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results