accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CSNCH3

CAS RN 857283-68-2

IUPAC Name: 2-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole
Synonyms: 2-methyl-4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3-thiazole
Molecular Weight (g/mol): 301.21
Molecular Formula: C16H20BNO2S
InChi Key: XCAUOIGPROQXRG-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results