accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 857283-71-7

IUPAC Name: tert-butyl 4-iodo-3,5-dimethyl-1H-pyrazole-1-carboxylate
Synonyms: tert-butyl 4-iodo-3,5-dimethylpyrazole-1-carboxylate
Molecular Weight (g/mol): 322.15
Molecular Formula: C10H15IN2O2
InChi Key: BUHPTZHNAXELNA-UHFFFAOYSA-N
SMILES: CC1=NN(C(=O)OC(C)(C)C)C(C)=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results