accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 857283-87-5

IUPAC Name: 1,4-dimethyl-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile
Synonyms: 1,4-dimethyl-2,3-dihydroquinoxaline-6-carbonitrile
Molecular Weight (g/mol): 187.25
Molecular Formula: C11H13N3
InChi Key: NBANRNWRIHAGBJ-UHFFFAOYSA-N
SMILES: CN1CCN(C)C2=C1C=CC(=C2)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results