accessibility menu, dialog, popup

UserName

Viewing profile as user:
NO

CAS RN 857283-89-7

IUPAC Name: 3-(pyrrolidin-1-yl)benzaldehyde
Synonyms: 3-(pyrrolidin-1-yl)benzaldehyde
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: KELYFFMWYRNGTF-UHFFFAOYSA-N
SMILES: O=CC1=CC(=CC=C1)N1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results