accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 857283-93-3

IUPAC Name: 4-(2-methyl-1,3-thiazol-4-yl)benzoyl chloride
Synonyms: 4-(2-methyl-1,3-thiazol-4-yl)benzoyl chloride
Molecular Weight (g/mol): 237.7
Molecular Formula: C11H8ClNOS
InChi Key: WJQGMUPBMHWAEX-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=C(C=C1)C(Cl)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results