accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 857284-20-9

IUPAC Name: tert-butyl 4-[5-(hydroxymethyl)pyridin-2-yl]piperazine-1-carboxylate
Synonyms: tert-butyl 4-[5-(hydroxymethyl)pyridin-2-yl]piperazine-1-carboxylate
Molecular Weight (g/mol): 293.37
Molecular Formula: C15H23N3O3
InChi Key: QKCHWJYIPFBVRD-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=NC=C(CO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results