accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 857284-26-5

IUPAC Name: methyl({2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}methyl)amine
Synonyms: methyl({2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-4-yl}methyl)amine
Molecular Weight (g/mol): 272.29
Molecular Formula: C12H11F3N2S
InChi Key: IBCDDBBGPGTKNC-UHFFFAOYSA-N
SMILES: CNCC1=CSC(=N1)C1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results