accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 857356-94-6

IUPAC Name: di-tert-butyl[3,4,5,6-tetramethyl-2',4',6'-tris(propan-2-yl)-[1,1'-biphenyl]-2-yl]phosphane
Synonyms: di-tert-butyl({2',4',6'-triisopropyl-3,4,5,6-tetramethyl-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol): 480.76
Molecular Formula: C33H53P
InChi Key: RCRYEYMHBHPZQD-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)C1=C(C)C(C)=C(C)C(C)=C1P(C(C)(C)C)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)