accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 857447-79-1

IUPAC Name: bis(tetrabutylazanium) hydrogen (hydrogen phosphonatooxy)phosphonate
Synonyms: bis(tetrabutylammonium ion) diphosphate(2-)
Molecular Weight (g/mol): 660.9
Molecular Formula: C32H74N2O7P2
InChi Key: WPGBCELIUCBRKN-UHFFFAOYSA-L
SMILES: OP([O-])(=O)OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results