accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 857530-80-4

IUPAC Name: 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular Weight (g/mol): 222.1
Molecular Formula: C11H19BN2O2
InChi Key: GNUDAJTUCJEBEI-UHFFFAOYSA-N
SMILES: CC1=C(B2OC(C)(C)C(C)(C)O2)C(C)=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results