CAS RN 85857-16-5
CAS RN 85857-16-5
IUPAC Name:
trimethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Synonyms:
trimethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
Molecular Weight (g/mol):
468.29
Molecular Formula:
C11H13F13O3Si
InChi Key:
BVQYIDJXNYHKRK-UHFFFAOYSA-N
SMILES:
CO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OC)OC
1
–
1
of
1
results
1
Trimethoxy(1H,1H,2H,2H-tridecafluoro-n-octyl)silane 97.0+%, TCI America™
CAS: 85857-16-5 Molecular Formula: C11H13F13O3Si Molecular Weight (g/mol): 468.286 MDL Number: MFCD00153261 InChI Key: BVQYIDJXNYHKRK-UHFFFAOYSA-N Synonym: Trimethoxy(1H,1H,2H,2H-perfluoro-n-octyl)silane PubChem CID: 2776334 IUPAC Name: trimethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane SMILES: CO[Si](CCC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(OC)OC