accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 85877-79-8

IUPAC Name: trimethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane
Synonyms: trimethoxy(3,3,4,4,5,5,6,6,6-nonafluorohexyl)silane
Molecular Weight (g/mol): 368.27
Molecular Formula: C9H13F9O3Si
InChi Key: IJROHELDTBDTPH-UHFFFAOYSA-N
SMILES: CO[Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(OC)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results