accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 858971-43-4

IUPAC Name: palladium(2+) [2-(benzenesulfinyl)ethanesulfinyl]benzene diacetate
Synonyms: white catalyst
Molecular Weight (g/mol): 502.89
Molecular Formula: C18H20O6PdS2
InChi Key: SNNYSJNYZJXIFE-UHFFFAOYNA-L
SMILES: [Pd++].CC([O-])=O.CC([O-])=O.O=S(CCS(=O)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)