accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 859-65-4

IUPAC Name: 1-phenyl-2-(triphenyl-λ⁵-phosphanylidene)ethan-1-one
Synonyms: 1-phenyl-2-(triphenyl-λ⁵-phosphanylidene)ethanone
Molecular Weight (g/mol): 380.43
Molecular Formula: C26H21OP
InChi Key: MZRSAJZDYIISJW-UHFFFAOYSA-N
SMILES: O=C(C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)