accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONO

CAS RN 85908-96-9

IUPAC Name: tert-butyl 2-oxopiperidine-1-carboxylate
Synonyms: tert-butyl 2-oxopiperidine-1-carboxylate
Molecular Weight (g/mol): 199.25
Molecular Formula: C10H17NO3
InChi Key: ULMHMJAEGZPQRY-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCCCC1=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)