accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 859169-20-3

IUPAC Name: ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
Synonyms: ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate
Molecular Weight (g/mol): 290.17
Molecular Formula: C16H23BO4
InChi Key: RPRBNRDIESZHFL-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results