CAS RN 85949-60-6
IUPAC Name:
yttrium(3+) triacetate tetrahydrate
Synonyms:
yttrium(3+) triacetate tetrahydrate
Molecular Weight (g/mol):
338.1
Molecular Formula:
C6H17O10Y
InChi Key:
AIQRTHPXPDTMBQ-UHFFFAOYSA-K
SMILES:
O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
1
–
2
of
2
results
1
Yttrium(III) acetate tetrahydrate, REacton™, 99.99% (REO)
CAS: 85949-60-6 Molecular Formula: C6H17O10Y Molecular Weight (g/mol): 338.10 MDL Number: MFCD00013048 InChI Key: AIQRTHPXPDTMBQ-UHFFFAOYSA-K IUPAC Name: yttrium(3+) triacetate tetrahydrate SMILES: O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
2
Yttrium(III) acetate tetrahydrate, 99.9% (REO)
CAS: 85949-60-6 Molecular Formula: C6H17O10Y Molecular Weight (g/mol): 338.10 MDL Number: MFCD00013048 InChI Key: AIQRTHPXPDTMBQ-UHFFFAOYSA-K IUPAC Name: yttrium(3+) triacetate tetrahydrate SMILES: O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O