CAS RN 85951-63-9
IUPAC Name:
N-{1-[2-(4-isothiocyanatophenyl)ethyl]piperidin-4-yl}-N-phenylpropanamide
Synonyms:
N-{1-[2-(4-isothiocyanatophenyl)ethyl]piperidin-4-yl}-N-phenylpropanamide
Molecular Weight (g/mol):
393.55
Molecular Formula:
C23H27N3OS
InChi Key:
VDKBIFPJULZUPU-UHFFFAOYSA-N
SMILES:
CCC(=O)N(C1CCN(CCC2=CC=C(C=C2)N=C=S)CC1)C1=CC=CC=C1
1
–
1
of
1
results
1
FIT, Tocris Bioscience™
CAS: 85951-63-9 Molecular Formula: C23H27N3OS Molecular Weight (g/mol): 393.549 InChI Key: VDKBIFPJULZUPU-UHFFFAOYSA-N Synonym: fentanyl isothiocyanate,fit,fentanyl ncs,n-1-2-4-isothiocyanatophenyl ethyl piperidin-4-yl-n-phenylpropanamide,fit alkylating agent,tocris-1480,n-1-2-4-isothiocyanatophenyl ethyl-4-piperidinyl-n-phenylpropanamide,n-1-2-4-isothiocyanato-phenyl-ethyl-piperidin-4-yl-n-phenyl-propionamide,propanamide, n-1-2-4-isothiocyanatophenyl ethyl-4-piperidinyl-n-phenyl PubChem CID: 84008 IUPAC Name: N-[1-[2-(4-isothiocyanatophenyl)ethyl]piperidin-4-yl]-N-phenylpropanamide SMILES: CCC(=O)N(C1CCN(CC1)CCC2=CC=C(C=C2)N=C=S)C3=CC=CC=C3