CAS RN 85959-83-7
IUPAC Name:
hafnium(4+) bis(pentamethylcyclopenta-1,3-dien-1-ide) dichloride
Synonyms:
hafnium(4+) bis(pentamethylcyclopenta-1,3-dien-1-ide) dichloride
Molecular Weight (g/mol):
519.85
Molecular Formula:
C20H30Cl2Hf
InChi Key:
YPRLSLCBPOTFGE-UHFFFAOYSA-L
SMILES:
[Cl-].[Cl-].[Hf+4].CC1=[C-]C(C)(C)C(C)=C1C.CC1=[C-]C(C)(C)C(C)=C1C
1
–
1
of
1
results
1
Promotions Available
Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride, 98+%
Bis(pentamethylcyclopentadienyl)hafnium(IV) dichloride, 98+%
CAS: 85959-83-7 Molecular Formula: C20H30Cl2Hf Molecular Weight (g/mol): 519.85 MDL Number: MFCD00058887 InChI Key: YPRLSLCBPOTFGE-UHFFFAOYSA-L Synonym: 1,2,3,5,5-pentamethylcyclopenta-1,3-diene;dichloride,hafnium,dichlorobis 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl PubChem CID: 21908586 IUPAC Name: hafnium(4+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene;dichloride SMILES: CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Cl-].[Cl-].[Hf+4]