accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 859833-12-8

IUPAC Name: 1-(6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl)methanamine hydrochloride
Synonyms: 1-(6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl)methanamine hydrochloride
Molecular Weight (g/mol): 219.64
Molecular Formula: C9H11ClFNO2
InChi Key: FBHYQVQESWEYTE-UHFFFAOYSA-N
SMILES: Cl.NCC1=C2OCOCC2=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results