accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNNHClHCl

CAS RN 859833-23-1

IUPAC Name: methyl({4-[(piperidin-1-yl)methyl]phenyl}methyl)amine dihydrochloride
Synonyms: methyl({[4-(piperidin-1-ylmethyl)phenyl]methyl})amine dihydrochloride
Molecular Weight (g/mol): 291.26
Molecular Formula: C14H24Cl2N2
InChi Key: VEBSTHXHQLTMJG-UHFFFAOYSA-N
SMILES: Cl.Cl.CNCC1=CC=C(CN2CCCCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results