accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 859850-65-0

IUPAC Name: methyl({[3-(piperidin-1-yl)phenyl]methyl})amine
Synonyms: methyl({[3-(piperidin-1-yl)phenyl]methyl})amine
Molecular Weight (g/mol): 204.32
Molecular Formula: C13H20N2
InChi Key: WHELHMLNRPDTJU-UHFFFAOYSA-N
SMILES: CNCC1=CC(=CC=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results