accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOBr

CAS RN 859851-04-0

IUPAC Name: 3-(2-bromophenyl)-5-methyl-1,2,4-oxadiazole
Synonyms: 3-(2-bromophenyl)-5-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 239.07
Molecular Formula: C9H7BrN2O
InChi Key: WLLDZYVHNSXWSY-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results