accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON(R)O(R)OPOOOPHOOO(R)O(R)(R)OH(S)HONNNH2NN(S)OH(R)HO

CAS RN 86-08-8

IUPAC Name: 3-acetyl-1-[(2R,3R,4S,5R)-5-{[({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]methyl}-3,4-dihydroxyoxolan-2-yl]-1λ⁵-pyridin-1-ylium
Synonyms: 3-acetyl-1-[(2R,3R,4S,5R)-5-[({[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy(hydroxy)phosphoryl phosphonato}oxy)methyl]-3,4-dihydroxyoxolan-2-yl]-1λ⁵-pyridin-1-ylium|APAD
Molecular Weight (g/mol): 662.44
Molecular Formula: C22H28N6O14P2
InChi Key: KPVQNXLUPNWQHM-RBEMOOQDSA-N
SMILES: CC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP(O)(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results