accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86-30-6

ONN

CAS RN 86-30-6

IUPAC Name: N-nitroso-N-phenylaniline
Synonyms: nitrosodiphenylamine delac J sconoc N-nitrosodifenylamin TJB vulcatard A vulcalent A redax difenylnitrosamin retarder J
Molecular Weight (g/mol): 198.23
Molecular Formula: C12H10N2O
InChi Key: UBUCNCOMADRQHX-UHFFFAOYSA-N
SMILES: O=NN(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results