accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86-38-4

IUPAC Name: 6,9-dichloro-2-methoxyacridine
Synonyms: 6,9-dichloro-2-methoxyacridine
Molecular Weight (g/mol): 278.13
Molecular Formula: C14H9Cl2NO
InChi Key: RYRNQWYNHLLOGX-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C3C=C(Cl)C=CC3=C(Cl)C2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)