accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOH

CAS RN 86-48-6

IUPAC Name: 1-hydroxynaphthalene-2-carboxylic acid
Synonyms: 1-hydroxy-2-naphthoic acid
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: SJJCQDRGABAVBB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=CC=CC2=C1O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (4)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)