accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86-49-7

IUPAC Name: 2-chloro-N,N-diethyl-4-nitroaniline
Synonyms: 2-chloro-N,N-diethyl-4-nitroaniline
Molecular Weight (g/mol): 228.68
Molecular Formula: C10H13ClN2O2
InChi Key: PCVJAZCSKCCKPQ-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(C=C1Cl)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results