accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 86-53-3

IUPAC Name: naphthalene-1-carbonitrile
Synonyms: 1-naphthalenecarbonitrile
Molecular Weight (g/mol): 153.18
Molecular Formula: C11H7N
InChi Key: YJMNOKOLADGBKA-UHFFFAOYSA-N
SMILES: N#CC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)