accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86-57-7

ONO

CAS RN 86-57-7

IUPAC Name: 1-nitronaphthalene
Synonyms: nitrol
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: RJKGJBPXVHTNJL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)