CAS RN 86-68-0
IUPAC Name:
6-methoxyquinoline-4-carboxylic acid
Synonyms:
quininic acid
Molecular Weight (g/mol):
203.2
Molecular Formula:
C11H9NO3
InChi Key:
XXLFLUJXWKXUGS-UHFFFAOYSA-N
SMILES:
COC1=CC=C2N=CC=C(C(O)=O)C2=C1
1
–
1
of
1
results
1
6-Methoxyquinoline-4-carboxylic Acid 98.0+%, TCI America™
CAS: 86-68-0 Molecular Formula: C11H9NO3 Molecular Weight (g/mol): 203.197 MDL Number: MFCD00024013 InChI Key: XXLFLUJXWKXUGS-UHFFFAOYSA-N Synonym: Quininic Acid PubChem CID: 345824 IUPAC Name: 6-methoxyquinoline-4-carboxylic acid SMILES: COC1=CC2=C(C=CN=C2C=C1)C(=O)O