accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86-81-7

IUPAC Name: 3,4,5-trimethoxybenzaldehyde
Synonyms: 3,4,5-trimethoxybenzaldehyde
Molecular Weight (g/mol): 196.2
Molecular Formula: C10H12O4
InChi Key: OPHQOIGEOHXOGX-UHFFFAOYSA-N
SMILES: COC1=CC(C=O)=CC(OC)=C1OC
Boiling Point 
  • (5)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (5)
  • (3)
Quantity 
  • (4)
  • (4)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (5)
  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (4)
  • (2)
  • (2)
  • (3)