accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNNHNN

CAS RN 86-93-1

IUPAC Name: 1-phenyl-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Synonyms: 1-phenyl-2H-1,2,3,4-tetrazole-5-thione
Molecular Weight (g/mol): 178.21
Molecular Formula: C7H6N4S
InChi Key: GGZHVNZHFYCSEV-UHFFFAOYSA-N
SMILES: S=C1N=NNN1C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)