accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNONH

CAS RN 86-96-4

IUPAC Name: 1,2,3,4-tetrahydroquinazoline-2,4-dione
Synonyms: 1,3-dihydroquinazoline-2,4-dione
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: SDQJTWBNWQABLE-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)