accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86-97-5

IUPAC Name: 5-aminonaphthalen-2-ol
Synonyms: 2-naphthalenol, 5-amino-
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: FSBRKZMSECKELY-UHFFFAOYSA-N
SMILES: NC1=C2C=CC(O)=CC2=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)