accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 86030-52-6

IUPAC Name: 2-{2-[2-amino-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido}acetic acid
Synonyms: {2-[2-amino-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido}acetic acid
Molecular Weight (g/mol): 337.38
Molecular Formula: C16H23N3O5
InChi Key: NWEGIYMHTZXVBP-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)C(N)CC1=CC=C(O)C=C1)C(=O)NCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)