CAS RN 86060-85-7
CAS RN 86060-85-7
IUPAC Name:
2,3,4,5,6-pentafluorophenyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetate
Synonyms:
2,3,4,5,6-pentafluorophenyl 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}acetate
Molecular Weight (g/mol):
463.36
Molecular Formula:
C23H14F5NO4
InChi Key:
LBSDTBJWUJIFBO-UHFFFAOYSA-N
SMILES:
FC1=C(F)C(F)=C(OC(=O)CNC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(F)=C1F
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Gly-OPfp, Advanced ChemTech
CAS: 86060-85-7 Molecular Formula: C23H14F5NO4 Molecular Weight (g/mol): 463.36 MDL Number: MFCD00037641 InChI Key: LBSDTBJWUJIFBO-UHFFFAOYSA-N IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)acetate SMILES: FC1=C(F)C(F)=C(OC(=O)CNC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(F)=C1F