accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)OOFFFFFHNOO

CAS RN 86060-87-9

IUPAC Name: 2,3,4,5,6-pentafluorophenyl (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylbutanoate
Synonyms: 2,3,4,5,6-pentafluorophenyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-methylbutanoate|FMOC-VAL-OPFP
Molecular Weight (g/mol): 505.44
Molecular Formula: C26H20F5NO4
InChi Key: TZEGAVSWQUEHAQ-QHCPKHFHSA-N
SMILES: CC(C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results